Learn More
TrueCare Biomedix Sterile Clean Room Gown, 52gsm+-3gsm Breathable PEPP, Level 3 impervious, CHEMO TESTED stand up collar with velcro closure in the back, with elastic wrists, seam seals, 1pcsterile bag,50pcsctn, XL
Catalog No. NC2217198
Encompass_Preferred
Sterile Clean Room Gown, 52gsm+-3gsm Breathable PEPP, Level 3 impervious, CHEMO TESTED stand up collar with velcro closure in the back, with elastic wrists, seam seals, 1pcsterile bag,50pcsctn, XL
This item is not returnable. View return policy
Catalog No. NC2217198 Supplier TrueCare Biomedix Supplier No. TCBA54STXL
Sterile Clean Room Gown, 52gsm+-3gsm Breathable PEPP, Level 3 impervious, CHEMO TESTED stand up collar with velcro closure in the back, with elastic wrists, seam seals, 1pcsterile bag,50pcsctn, XL