Learn More
Poltex Clean Room Liquid Resistant PETG Logbook Sleeve, Tape Mount
SDP
Catalog No. 22022573
Change view
Click to view available options
Cover Color:
Clear
Catalog No. Cover Color
22-022-573 Clear
1 options
Catalog No. 22-022-573 Supplier Poltex Supplier No. DRYLOGT
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Suitable for clean room applications, the bottom keeper slides to the side for document insertion. Top and sides are sealed, bottom slides into internal channels to keep inside document dry when subject to liquids. Tape mount (VHB tape included)
  • PETG logbook holder
  • Suitable for clean room applications
  • Bottom keeper slides to the side for document insertion
  • Top and sides are sealed, bottom slides into internal channels to keep inside document dry when subject to liquids
  • Mounts with VHB Tape
Specifications
Material Plastic
Ruled No
Color Clear
Cover Color Clear
Length (Metric) 304.80 mm
Width (Metric) 228.60 mm
Autoclavable Not Autoclavable
Disposable Reusable
Plain paper Note paper
Product Type Logbook Sleeve
Sterility Non-sterile