Learn More
Poltex Clean Room Liquid Resistant Sign Sleeve
SDP
Catalog No. 22000682
Change view
Click to view available options
Material:
Polycarbonate
Catalog No. Material
22-000-682 Polycarbonate
1 options
This item is not returnable. View return policy
Catalog No. 22-000-682 Supplier Poltex Supplier No. DRYSIGT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Poltex Liquid Resistant Sign Sleeve is suitable for clean room applications.
PETG sign sleeve. Suitable for clean room applications. Bottom keeper slides to the side for document insertion. Top and sides are sealed, bottom slides into internal channels to keep inside document dry when subject to liquids. Mounts with VHB Tape.
Specifications
Material Polycarbonate
Size (English) 8.5 x 11 in.
Mounting Type Self-adhesive
Shape Rectangular
Languages Universal
Color Clear