Learn More
MSC PRO-SAFE Clean Room Mat, Tacky Sheets, 36 x 45 in, 30 Layers
Catalog No. 502835330
Encompass
This clean room mat consists of tacky sheets designed to capture contaminants before entering a cleanroom environment. Each mat features 30 layers of adhesive film to effectively remove particles from shoes and cart wheels. It measures 36 in wide by 45 in long and comes in blue.
  • Tacky sheets surface material
  • 30 layers per mat for extended use
  • 36 in wide by 45 in long dimensions
  • Smooth surface pattern
  • Suitable for dry environments
  • Non-reusable design
Catalog No. 50-283-5330 Supplier MSC Supplier No. 53136891
This clean room mat consists of tacky sheets designed to capture contaminants before entering a cleanroom environment. Each mat features 30 layers of adhesive film to effectively remove particles from shoes and cart wheels. It measures 36 in wide by 45 in long and comes in blue.
  • Tacky sheets surface material
  • 30 layers per mat for extended use
  • 36 in wide by 45 in long dimensions
  • Smooth surface pattern
  • Suitable for dry environments
  • Non-reusable design