Learn More
MSC Armstrong World Ceiling Tiles, Mineral Fiber, 48 x 24 inch, Class A Fire Rating
Catalog No. 502836159
Encompass
These ceiling tiles are designed for use in clean rooms and food processing areas, providing durability with washability, scrubability, water repellent, and soil resistance. They offer resistance to mold, mildew, and sag, and provide fire-resistant properties when installed in applicable UL assemblies.
  • Wet-formed mineral fiber material
  • Dimensions: 48 x 24 inch
  • Class A fire rating
  • Sag, mold, and mildew resistant
  • Washable, scrubbable, water repellent, and soil resistant
  • GreenGuard Gold Certified
  • Ideal for clean rooms and food processing areas
  • Light reflectance of 80%
  • 41% recycled content
Catalog No. 50-283-6159 Supplier MSC Supplier No. 89294086
These ceiling tiles are designed for use in clean rooms and food processing areas, providing durability with washability, scrubability, water repellent, and soil resistance. They offer resistance to mold, mildew, and sag, and provide fire-resistant properties when installed in applicable UL assemblies.
  • Wet-formed mineral fiber material
  • Dimensions: 48 x 24 inch
  • Class A fire rating
  • Sag, mold, and mildew resistant
  • Washable, scrubbable, water repellent, and soil resistant
  • GreenGuard Gold Certified
  • Ideal for clean rooms and food processing areas
  • Light reflectance of 80%
  • 41% recycled content