Learn More
MSC Nilfisk Cleanroom Vacuum, Electric, 3.25 gal, ULPA Filter, 1 ea
Catalog No. 502868913
Encompass
The Nilfisk IVT 1000CR cleanroom vacuum is engineered for demanding cleanroom environments, featuring a multistage filtration system and quiet operation for critical applications. Its robust stainless steel construction ensures durability and compatibility with stringent sterilization protocols.
  • Multistage filtration system
  • ULPA exhaust filter standard
  • Compatible with ISO 4 (Class 10) cleanrooms
  • AISI 316 stainless steel construction
  • Trolley and container are autoclavable
  • Quiet operation at 61 dB(A)
  • Compact and modular design for easy maintenance
  • Offers 3.25 gal dry collection capacity
Catalog No. 50-286-8913 Supplier MSC Supplier No. 35210392
The Nilfisk IVT 1000CR cleanroom vacuum is engineered for demanding cleanroom environments, featuring a multistage filtration system and quiet operation for critical applications. Its robust stainless steel construction ensures durability and compatibility with stringent sterilization protocols.
  • Multistage filtration system
  • ULPA exhaust filter standard
  • Compatible with ISO 4 (Class 10) cleanrooms
  • AISI 316 stainless steel construction
  • Trolley and container are autoclavable
  • Quiet operation at 61 dB(A)
  • Compact and modular design for easy maintenance
  • Offers 3.25 gal dry collection capacity