Learn More
Poltex™ Clean Room Wipe Dispenser Wall Mount
SDP
Catalog No. 22000766
Change view
Click to view available options
Mounting:
Freestanding or Wall Mount
Catalog No. Mounting
22-000-766 Freestanding or Wall Mount
1 options
Catalog No. 22-000-766 Supplier Poltex™ Supplier No. CLNWIPW
Add to Cart
Add to Cart
Cleanroom wipe holder with lid that mounts to wall.
  • Holds wipes for clean room
  • Made of durable PETG construction
  • Cleanable
  • Comes with rubber bumpons on base for non-slip placement
  • Comes with wall mount hardware
Specifications
Product Type Wipe Dispenser
Color Clear
Dimensions (D x W x H) Interior 10 x 10.5 x 5.5 in.
Holds Wipes
Material PETG
Mounting Freestanding or Wall Mount
No. of Compartments 1
Product Style Hinged Lid