Learn More
UCT LLC Clean Screen Xcel I 130mg 10mL
SDP
Catalog No. NC1763347
Encompass
Clean Screen Xcel solid phase extraction columns are designed to reduce the number of steps in an extraction. The result is a column that reduces sample prep times and minimizes the quantity of solvent required. Additional advantages include reduced sample size and improved cleanliness and recovery.
Catalog No. NC1763347 Supplier UCT LLC Supplier No. ZSXCE010
Clean Screen Xcel solid phase extraction columns are designed to reduce the number of steps in an extraction. The result is a column that reduces sample prep times and minimizes the quantity of solvent required. Additional advantages include reduced sample size and improved cleanliness and recovery.