Learn More
MSC CLEAN / SHINE PAD
Catalog No. 5000787963
Encompass
Scotch-Brite™ Clean & Shine Pads are engineered for floor cleaning and polishing, allowing users to clean and shine floors simultaneously with a low-speed scrubber. They reduce the need for burnishing, saving costs and labor.
  • Cleans and shines floors in a single step
  • Reduces or eliminates the need for burnishing
  • Removes black marks three times faster than traditional cleaning pads
  • Two-sided for extended life
  • Effective on most coated and uncoated hard floors with water or a neutral cleaner
  • Improves gloss with each use
  • Features a contrasting color for visible wear indication
Catalog No. 50-007-87963 Supplier MSC Supplier No. 97588214
Scotch-Brite™ Clean & Shine Pads are engineered for floor cleaning and polishing, allowing users to clean and shine floors simultaneously with a low-speed scrubber. They reduce the need for burnishing, saving costs and labor.
  • Cleans and shines floors in a single step
  • Reduces or eliminates the need for burnishing
  • Removes black marks three times faster than traditional cleaning pads
  • Two-sided for extended life
  • Effective on most coated and uncoated hard floors with water or a neutral cleaner
  • Improves gloss with each use
  • Features a contrasting color for visible wear indication