Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOTRAX™ Clean-Step™ Frame
Shop All NoTrax Products
Click to view available options
Length (English):
20 in.
26 in.
28 in.
Width (English):
38 in.
47 in.
Specifications
Specifications
For Use With (Equipment) | For Clean-Step™ Tacky Mat |
Length (English) | 20 in. |
Quantity | 1 Ea. |
Width (English) | 38 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction