Learn More
Nelson Jameson Company CLEAN TRACE SURFACE PROTN SWAB
Supplier:  Nelson Jameson Company 3513062 C
Encompass
3M™ Clean-Trace™ Surface Protein (Allergen) Test Swab ALLTEC60, 60 per case. Detects as little as 3 µg of protein on surfaces and in hard to reach areas. Provides a tool to help assess and improve hygienic performance. Visual reading of a color change indicates the level of cleanliness. Real time results allow immediate corrective action. Recommended Industry - Food Services & Catering, Fruits & Vegetables, Meat, Pet Food & Animal Feed, Poultry & Eggs, Prepared & Processed Foods, Seafood. Test Type Environmental
Catalog No. NC0901553