Learn More
Norgen Biotek Corp RNA Clean-Up and Concentration Kit, 96-well plate format, RNA Input up to 50 μg, Elution Volume 75 μL, Spin column chromatography, For total RNA including miRNA
Catalog No. NC3350814
Encompass_Preferred
RNA Clean-Up and Concentration Kit, 96-well plate format, RNA Input up to 50 μg, Elution Volume 75 μL, Spin column chromatography, For total RNA including miRNA
Catalog No. NC3350814 Supplier Norgen Biotek Corp Supplier No. 25100
RNA Clean-Up and Concentration Kit, 96-well plate format, RNA Input up to 50 μg, Elution Volume 75 μL, Spin column chromatography, For total RNA including miRNA