Learn More
Bio Rad Laboratories CLEAN UP KIT 2D
Catalog No. NC9099635
Encompass
ReadyPrep 2-D Cleanup Kit, Sufficient to Process 50 Protein Samples, 1 to 100microliter Each, Procedure TCA-Like Precipitation to Remove Salts, Detergents, Lipids, Phenolic Compounds
Catalog No. NC9099635 Supplier Bio Rad Laboratories Supplier No. 1632130
ReadyPrep 2-D Cleanup Kit, Sufficient to Process 50 Protein Samples, 1 to 100microliter Each, Procedure TCA-Like Precipitation to Remove Salts, Detergents, Lipids, Phenolic Compounds