Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Clean-Walk Mat

Catalog No. 17623855 Shop All 3M Products
Change view
Click to view available options
Color:
White
Dimensions (L x W):
36 x 18 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Dimensions (L x W)
17-623-855 White 36 x 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-855 Supplier 3M™ Supplier No. 7000049618

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ Clean Walk Mat 5830 is a multiple layer sticky mat with individually tabbed layers.

  • Thin floor mat profile minimizes tripping hazard. Bottom layer of the pad firmly attaches to most smooth, clean surfaces, such as tile or sealed concrete.
  • Sticky mat captures dirt on contact before tracking it from one area to another. It prolongs life and appearance of floors and carpet and reduces facility maintenance and clean up. Pad length is sufficient for two step contact.
  • Multiple adhesive layers allow for easily renewed surface. Individually numbered tabs ensure that only one layer is removed at a time.
  • Facilities, general industrial, maintenance, military, aerospace, construction, transportation, truck, bus, commercial vehicle, manufacturing, automotive, RV, ship yards, metal working, appliance, electronics, food and beverage.
  • Protecting, preventing, prolonging life, controlling contamination, reducing dirt and dust, keeping clean, removing dirt
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 36 x 18 in.
Type Entrance Mat
Quantity 4/Cs.
Thickness (English) 0.055 in.
Color White
Length (English) 36 in.
Length (Metric) 914 mm
Material Acrylic, Polyethylene
Width (English) 18 in.
Width (Metric) 457 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.