Learn More
Hach Company Clean Water Analog ORP Sensor
Catalog No. 5000207633 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - Clean Water Analog ORP SensorClean Water Online Process ORP Sensor - Platinum ORP Electrode PPS Housing Convertible Mount 4 5m Cable Analog Cable Connection
Catalog No. 50-002-07633 Supplier Hach Company Supplier No. RD1R5
Hach Online Process ORP Sensor - Clean Water Analog ORP SensorClean Water Online Process ORP Sensor - Platinum ORP Electrode PPS Housing Convertible Mount 4 5m Cable Analog Cable Connection