Learn More
Norgen Biotek Corp CleanAll DNA/RNA Clean-Up and Concentration Micro Kit
Catalog No. NC1508300
Encompass_Preferred
Norgen's CleanAll DNA/RNA Clean-Up and Concentration Micro Kit provides a rapid method for the purification, cleanup and concentration of RNA or DNA from different isolation methods or upstream applications. The kit can be used as an alternative to organic extraction and ethanol precipitation to clean up various enzymatic reactions. The kit purifies RNA from phenol/guanidine-based protocols or from various upstream enzymatic reactions such as DNase treatment, labeling and in vitro transcription. Endotoxins can be removed from previously purified RNA solutions down to 0.1 EU/µg DNA or less. The kit can be used to clean up DNA from digestions, ligations, PCR reactions, DNA modification reactions and staining.Purify all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA and small interfering RNA.Purify all sizes of DNA, from small PCR products, native or linearized plasmids, oligonucleotides, to genomic DNA.
Catalog No. NC1508300 Supplier Norgen Biotek Corp Supplier No. 23800
Norgen's CleanAll DNA/RNA Clean-Up and Concentration Micro Kit provides a rapid method for the purification, cleanup and concentration of RNA or DNA from different isolation methods or upstream applications. The kit can be used as an alternative to organic extraction and ethanol precipitation to clean up various enzymatic reactions. The kit purifies RNA from phenol/guanidine-based protocols or from various upstream enzymatic reactions such as DNase treatment, labeling and in vitro transcription. Endotoxins can be removed from previously purified RNA solutions down to 0.1 EU/µg DNA or less. The kit can be used to clean up DNA from digestions, ligations, PCR reactions, DNA modification reactions and staining.Purify all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA and small interfering RNA.Purify all sizes of DNA, from small PCR products, native or linearized plasmids, oligonucleotides, to genomic DNA.