Learn More
Trilink Bio Technologies Inc CleanCap Cas9 mRNA (5moU), 1mg
SDP
Catalog No. NC1692492
Encompass
Purity Passes Agarose Gel Mobility, Cas9 mRNA expresses a version of the Streptococcus pyogenes SF370 Cas9 protein (CRISPR Associated Protein 9). Cas9 functions as part of the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) genome editing system. In the CRISPR system, an RNA guide sequence targets the site of interest and the Cas9 protein is employed to perform double stranded DNA cleavage.
Catalog No. NC1692492 Supplier Trilink Bio Technologies Inc Supplier No. L72061000
Purity Passes Agarose Gel Mobility, Cas9 mRNA expresses a version of the Streptococcus pyogenes SF370 Cas9 protein (CRISPR Associated Protein 9). Cas9 functions as part of the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) genome editing system. In the CRISPR system, an RNA guide sequence targets the site of interest and the Cas9 protein is employed to perform double stranded DNA cleavage.