Learn More
Trilink Bio Technologies Inc CLEANCAP CRE MRNA (5MOU) 5X1MG
SDP
Catalog No. NC2019358
Encompass
NLS-Cre Recombinase mRNA is a capped and polyadenylated messenger RNA encoding Cre recombinase fused to a nuclear localization sequence (NLS). Cre recombinase is a tyrosine recombinase that catalyzes recombination between two loxP sites.This mRNA is capped using CleanCap, TriLink's proprietary co-transciptional capping method, which results in the naturally occuring Cap 1 structure with high capping efficiency. It is polyadenylated, modified with 5-methoxyuridine and optimized for mammalian systems. It mimics a fully processed mature mRNA.
Catalog No. NC2019358 Supplier Trilink Bio Technologies Inc Supplier No. L72115
Add to Cart
Add to Cart
Request a Quote
NLS-Cre Recombinase mRNA is a capped and polyadenylated messenger RNA encoding Cre recombinase fused to a nuclear localization sequence (NLS). Cre recombinase is a tyrosine recombinase that catalyzes recombination between two loxP sites.This mRNA is capped using CleanCap, TriLink's proprietary co-transciptional capping method, which results in the naturally occuring Cap 1 structure with high capping efficiency. It is polyadenylated, modified with 5-methoxyuridine and optimized for mammalian systems. It mimics a fully processed mature mRNA.