Learn More
Trilink Bio Technologies Inc CleanCap EGFP mRNA (5moU), 1 mg
SDP
Catalog No. NC1411424
Encompass
The EGFP mRNA will express an enhanced version of the green fluorescent protein, originally isolated from the jellyfish, Aequorea victoria. EGFP is a commonly used direct detection reporter in mammalian cell culture, yielding bright green fluorescence with an emission peak at 509 nm.
Catalog No. NC1411424 Supplier Trilink Bio Technologies Inc Supplier No. L72011000
The EGFP mRNA will express an enhanced version of the green fluorescent protein, originally isolated from the jellyfish, Aequorea victoria. EGFP is a commonly used direct detection reporter in mammalian cell culture, yielding bright green fluorescence with an emission peak at 509 nm.