Learn More
Trilink Bio Technologies Inc CleanCap EGFP mRNA (5moU), 100 ug
SDP
Catalog No. NC1254220
Encompass
The EGFP mRNA will express an enhanced version of the green fluorescent protein, originally isolated from the jellyfish, Aequorea victoria. EGFP is a commonly used direct detection reporter in mammalian cell culture, yielding bright green fluorescence with an emission peak at 509 nm.
Catalog No. NC1254220 Supplier Trilink Bio Technologies Inc Supplier No. L7201100
The EGFP mRNA will express an enhanced version of the green fluorescent protein, originally isolated from the jellyfish, Aequorea victoria. EGFP is a commonly used direct detection reporter in mammalian cell culture, yielding bright green fluorescence with an emission peak at 509 nm.