Learn More
Trilink Bio Technologies Inc CleanCap Reagent AG, 5 µmole
SDP
Catalog No. NC1581351
Encompass
Molecular Formula C32H43N15O24P4 (free acid), Purity ≥95% by AX-HPLC, Molecular Weight 1145.60 g/mole (free acid), CleanCap Reagent AG is TriLink’s proprietary co-transcriptional capping reagent for in vitro transcription of 5’ capped mRNA resulting in a Cap 1 structure. CleanCap has shown to provide up to 98% capping efficiency1. PLEASE NOTE: TriLink recommends using CleanCap AG whenever possible. CleanCap AG requires an AG initiator. CleanCap AG typically provides >90% capped material generating a naturally occurring Cap 1 structure. Unlike CleanCap AG, using CleanCap GG (cat. no. N-7133 or N-7433) may result in comparable ARCA transcriptional stuttering and yield.
Catalog No. NC1581351 Supplier Trilink Bio Technologies Inc Supplier No. N71135
Molecular Formula C32H43N15O24P4 (free acid), Purity ≥95% by AX-HPLC, Molecular Weight 1145.60 g/mole (free acid), CleanCap Reagent AG is TriLink’s proprietary co-transcriptional capping reagent for in vitro transcription of 5’ capped mRNA resulting in a Cap 1 structure. CleanCap has shown to provide up to 98% capping efficiency1. PLEASE NOTE: TriLink recommends using CleanCap AG whenever possible. CleanCap AG requires an AG initiator. CleanCap AG typically provides >90% capped material generating a naturally occurring Cap 1 structure. Unlike CleanCap AG, using CleanCap GG (cat. no. N-7133 or N-7433) may result in comparable ARCA transcriptional stuttering and yield.