Learn More
Bulldog Bio Inc CleanDTR Dye Terminator Removal Beads, 500ml
SDP
Catalog No. NC1933446
Encompass
CleanDTR is an efficient, paramagnetic bead-based system, designed to remove unincorporated dye terminators from Sanger sequencing reactions prior to capillary electrophoresis. The process involves just three simple steps: bind, wash, and elute.
Catalog No. NC1933446 Supplier Bulldog Bio Inc Supplier No. CDTR500
Add to Cart
Add to Cart
Request a Quote
CleanDTR is an efficient, paramagnetic bead-based system, designed to remove unincorporated dye terminators from Sanger sequencing reactions prior to capillary electrophoresis. The process involves just three simple steps: bind, wash, and elute.