Learn More
Bulldog Bio Inc CleanDTR Dye Terminator Removal Beads, 50ml
SDP
Catalog No. NC1933444
Encompass_Preferred
CleanDTR is an efficient, paramagnetic bead-based system, designed to remove unincorporated dye terminators from Sanger sequencing reactions prior to capillary electrophoresis. The process involves just three simple steps: bind, wash, and elute.
Catalog No. NC1933444 Supplier Bulldog Bio Inc Supplier No. CDTR050
CleanDTR is an efficient, paramagnetic bead-based system, designed to remove unincorporated dye terminators from Sanger sequencing reactions prior to capillary electrophoresis. The process involves just three simple steps: bind, wash, and elute.