Learn More
Bulldog Bio Inc CleanDTR Dye Terminator Removal Beads, 5ml
SDP
Catalog No. NC1933443
Encompass
CleanDTR is an efficient, paramagnetic bead-based system, designed to remove unincorporated dye terminators from Sanger sequencing reactions prior to capillary electrophoresis. The process involves just three simple steps: bind, wash, and elute.
Catalog No. NC1933443 Supplier Bulldog Bio Inc Supplier No. CDTR005
CleanDTR is an efficient, paramagnetic bead-based system, designed to remove unincorporated dye terminators from Sanger sequencing reactions prior to capillary electrophoresis. The process involves just three simple steps: bind, wash, and elute.