Learn More
Roche Diagnostics CLEANER ALKALINE BIO
Supplier:  Roche Diagnostics 06689485001
Encompass
This product is used specifically with the IgG application to prevent sample carry over from high to low concentration IgG.
Catalog No. 50-589-444