This liquid disinfectant cleaner utilizes a non-abrasive formula that combines heavy-duty detergents with the stain-removal power of bleach for cleaning, sanitizing, deodorizing, and disinfecting surfaces. It is suitable for restroom, kitchen, and other hard-surface applications including fiberglass, glass, grout, metal, plastic, vinyl, and sealed or engineered stone.
- Cleans greasy soap scum with 50% fewer strokes.
- Kills 99.9% of bacteria and viruses in one minute when used according to label instructions.
- Functions as a hospital-use disinfectant effective against pathogens including Clostridium difficile spores, Staphylococcus aureus, MRSA, and Norovirus.
- Features a fresh scent.
- Complies with Title 17, California Code of Regulations maximum allowable V.O.C. content for general purpose cleaners.
- EPA registration number 3573-77.