Learn More
ACL Inc Mat & Table Top Cleaner, 1 quart, 12/cs
Catalog No. 501037901
Encompass_Preferred
Cleans, rejuvenates, and extends the life of ESD mats. Designed for cleaning ANY Production area, including assembly, testing and areas where sensitive electronics components are handled. Contains no alkali or ammonia.
Catalog No. 50-103-7901 Supplier ACL Inc Supplier No. 6001
Cleans, rejuvenates, and extends the life of ESD mats. Designed for cleaning ANY Production area, including assembly, testing and areas where sensitive electronics components are handled. Contains no alkali or ammonia.