Learn More
MSC Multi-Clean Bio Power Plus Enzyme Cleaner, Liquid, 1 gal
Catalog No. 502866621
Encompass
Bio Power Plus is a water-based enzyme cleaner that is classified as non-hazardous, stable, and readily biodegradable. This liquid cleaner is completely soluble in water and has a spice odor.
  • Classified as non-hazardous
  • Stable and readily biodegradable
  • Liquid physical state
  • White appearance with a spice odor
  • Has a pH between 6 and 8
  • Non-flammable with a boiling point > 100°C
  • Contains viable bacterial cultures and other functional ingredients
  • Container size: 1 gal
Catalog No. 50-286-6621 Supplier MSC Supplier No. 73061657
Bio Power Plus is a water-based enzyme cleaner that is classified as non-hazardous, stable, and readily biodegradable. This liquid cleaner is completely soluble in water and has a spice odor.
  • Classified as non-hazardous
  • Stable and readily biodegradable
  • Liquid physical state
  • White appearance with a spice odor
  • Has a pH between 6 and 8
  • Non-flammable with a boiling point > 100°C
  • Contains viable bacterial cultures and other functional ingredients
  • Container size: 1 gal