Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Autotech Mfg. Industrial Cleaner/Degreaser, V.O.C. Compliant, Pine Scent, 1 gal

Catalog No. 502839030
Encompass

This industrial cleaner/degreaser is a V.O.C. compliant, non-flammable, general purpose detergent. It features a distinctive pink appearance and a refreshing pine scent, making it suitable for various cleaning tasks.

  • V.O.C. compliant formula.
  • Non-flammable liquid.
  • Pine scent for a pleasant cleaning experience.
  • General purpose detergent for diverse applications.
  • Distinctive pink appearance.
  • Causes skin irritation.
  • Causes eye irritation.
  • May cause respiratory irritation.
  • pH range of 12.0-13.5.
  • Specific gravity of 1.00-1.07.

Catalog No. 50-283-9030 Supplier MSC Supplier No. 53023982
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This industrial cleaner/degreaser is a V.O.C. compliant, non-flammable, general purpose detergent. It features a distinctive pink appearance and a refreshing pine scent, making it suitable for various cleaning tasks.

  • V.O.C. compliant formula.
  • Non-flammable liquid.
  • Pine scent for a pleasant cleaning experience.
  • General purpose detergent for diverse applications.
  • Distinctive pink appearance.
  • Causes skin irritation.
  • Causes eye irritation.
  • May cause respiratory irritation.
  • pH range of 12.0-13.5.
  • Specific gravity of 1.00-1.07.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.