Learn More
Med Vet International Hibiclens Surgical Scrub, 4-ounce pump bottle, Non-Sterile, 57541, Each
Catalog No. 503044798
Encompass_Preferred
A surgical scrub, patient pre-op skin preparation, healthcare personnel hand wash, and a skin wound and general skin cleaner A trusted leader in antimicrobial cleaners, bonds with the skin to create a protective, germ-killing field In addition to the fast acting antimicrobial effect, HIBICLENS' non-drying, low skin irritation potential promotes compliance with the frequent hand washing associated with today's clinical protocols Clear liquid, pink color, and fragranced
Catalog No. 50-304-4798 Supplier Med Vet International Supplier No. 1128148EA
A surgical scrub, patient pre-op skin preparation, healthcare personnel hand wash, and a skin wound and general skin cleaner A trusted leader in antimicrobial cleaners, bonds with the skin to create a protective, germ-killing field In addition to the fast acting antimicrobial effect, HIBICLENS' non-drying, low skin irritation potential promotes compliance with the frequent hand washing associated with today's clinical protocols Clear liquid, pink color, and fragranced