Learn More
Siemens Healthineers Cleaner SCS
Catalog No. 23044679 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
6 x 5 mL
Catalog No. Quantity
23-044-679 6 x 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-679 Supplier Siemens Healthineers Supplier No. 10446136
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cleaning solution for BN II, BN ProSpec™ and BCS™ Systems.
TRUSTED_SUSTAINABILITY
Specifications
Type Cleaner
For Use With (Equipment) BN II, BN ProSpec™ & BCS™ Systems
Quantity 6 x 5 mL