Learn More
Bj Long Pipe Cleaner Brushes, 33mm, 50ft per roll, 6 rolls per case
Catalog No. NC2189529
Encompass
This item has a minimum order quantity of 3 cases per supplier requirements.
Cut at any length for cleaning small pipets, stopcocks, capillary tubes. May be used for slightly larger sizes by bending in moderate zig-zag pattern
Catalog No. NC2189529 Supplier Bj Long Supplier No. 03642A
This item has a minimum order quantity of 3 cases per supplier requirements.
Cut at any length for cleaning small pipets, stopcocks, capillary tubes. May be used for slightly larger sizes by bending in moderate zig-zag pattern