Learn More
BRANSON ULTRASONICS CORPORATION (3P) CLEANER ULTRASONIC B200 117V
Catalog No. NC3721522
Encompass_Preferred
Branson Ultrasonics Bransonic Ultrasonic Cleaner Model B200, Voltage: 120V, Capacity: 400mL (15 oz.), Hertz: 60Hz, Model: B200, Depth Tank: 8.8cm, Electrical Requirements: 120V 60Hz
Catalog No. NC3721522 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 100951010
Branson Ultrasonics Bransonic Ultrasonic Cleaner Model B200, Voltage: 120V, Capacity: 400mL (15 oz.), Hertz: 60Hz, Model: B200, Depth Tank: 8.8cm, Electrical Requirements: 120V 60Hz