Learn More
MSC Detco Phosphoric Acid Cleaner, 1 gal, Stainless Steel, 4/Case
Catalog No. 502816312
Encompass
Phos-Master is a phosphoric acid cleaner designed for effective cleaning in food and beverage plants, specifically formulated for stainless steel surfaces.
  • All-purpose cleaner
  • Liquid formula
  • Supplied in 1 gal bottles
  • Unscented
  • Suitable for food and beverage plants
  • Not a disinfectant
  • Specifically designed for stainless steel
  • Detergent-based formula
  • Dilutable concentrate
  • Biodegradable
  • Low 1% VOC content
Catalog No. 50-281-6312 Supplier MSC Supplier No. 33702101
Phos-Master is a phosphoric acid cleaner designed for effective cleaning in food and beverage plants, specifically formulated for stainless steel surfaces.
  • All-purpose cleaner
  • Liquid formula
  • Supplied in 1 gal bottles
  • Unscented
  • Suitable for food and beverage plants
  • Not a disinfectant
  • Specifically designed for stainless steel
  • Detergent-based formula
  • Dilutable concentrate
  • Biodegradable
  • Low 1% VOC content