Learn More
MSC Rite-Kem All-Purpose Cleaner & Degreaser, 1 gal, Liquid, Citrus Based
Catalog No. 502839174
Encompass
This citrus-based cleaner and degreaser is a fast-acting compound formulated to effectively remove grease, oil, ink, fats, tar, adhesives, and other soils. It is USDA C1 compliant.
  • Liquid form
  • 1 gal bottle
  • Pleasant scent
  • Not a disinfectant
  • Biodegradable, water-based formula
  • Dilutable concentrate solution
  • Non-acidic
  • VOC content of 5%
Catalog No. 50-283-9174 Supplier MSC Supplier No. 48175962
This citrus-based cleaner and degreaser is a fast-acting compound formulated to effectively remove grease, oil, ink, fats, tar, adhesives, and other soils. It is USDA C1 compliant.
  • Liquid form
  • 1 gal bottle
  • Pleasant scent
  • Not a disinfectant
  • Biodegradable, water-based formula
  • Dilutable concentrate solution
  • Non-acidic
  • VOC content of 5%