Learn More
MSC Zep Multi-Surface Cleaner Concentrate, Extra Heavy-Duty, Liquid, 275 gal
Catalog No. 502818590
Encompass
Zep A-One is a concentrated, water-based, low-foam industrial cleaner designed for tough cleaning tasks. This versatile formula effectively cleans concrete, masonry, brick, tile, and industrial floors by suspending soils and brightening surfaces while removing grease, oil, and light rust stains. It is ideal for use in automatic scrubbers and various industrial applications, providing efficient cleaning with a pleasant, clean fragrance.
  • Removes stubborn dirt, salts, carbon, and light rust stains
  • Cleans concrete floors, tiles, brick, and masonry
  • Lifts and suspends soils, grease, and light rust stains, leaving surfaces cleaner and brighter
  • Suitable for mop, brush, spray, or automatic scrubber application
  • Concentrated, water-based, low-foam formula
  • Meets USDA and NSF A4 standards
Catalog No. 50-281-8590 Supplier MSC Supplier No. 17643545
Add to Cart
Add to Cart
Request a Quote
Zep A-One is a concentrated, water-based, low-foam industrial cleaner designed for tough cleaning tasks. This versatile formula effectively cleans concrete, masonry, brick, tile, and industrial floors by suspending soils and brightening surfaces while removing grease, oil, and light rust stains. It is ideal for use in automatic scrubbers and various industrial applications, providing efficient cleaning with a pleasant, clean fragrance.
  • Removes stubborn dirt, salts, carbon, and light rust stains
  • Cleans concrete floors, tiles, brick, and masonry
  • Lifts and suspends soils, grease, and light rust stains, leaving surfaces cleaner and brighter
  • Suitable for mop, brush, spray, or automatic scrubber application
  • Concentrated, water-based, low-foam formula
  • Meets USDA and NSF A4 standards