Learn More
MSC Rite-Kem All-Purpose Cleaner & Degreaser, Liquid, 55 gal, Biodegradable
Catalog No. 502837614
Encompass
This concentrated, biodegradable, and water-based all-purpose cleaner and degreaser is formulated with emulsifiers, dispersants, and penetrants. It effectively attacks and breaks down grease and grime from various surfaces, including plastic, rubber, and metal, allowing for easy rinsing. It is designed as a dilutable concentrate and comes in a 55-gallon plastic drum.
  • Concentrated liquid formula.
  • Biodegradable and water-based.
  • Contains emulsifiers, dispersants, and penetrants.
  • Effectively removes grease and grime.
  • Suitable for plastic, rubber, and metal surfaces.
  • Allows for easy rinsing.
  • Dilutable for various cleaning tasks.
  • Comes in a large 55-gallon drum.
Catalog No. 50-283-7614 Supplier MSC Supplier No. 48175657
This concentrated, biodegradable, and water-based all-purpose cleaner and degreaser is formulated with emulsifiers, dispersants, and penetrants. It effectively attacks and breaks down grease and grime from various surfaces, including plastic, rubber, and metal, allowing for easy rinsing. It is designed as a dilutable concentrate and comes in a 55-gallon plastic drum.
  • Concentrated liquid formula.
  • Biodegradable and water-based.
  • Contains emulsifiers, dispersants, and penetrants.
  • Effectively removes grease and grime.
  • Suitable for plastic, rubber, and metal surfaces.
  • Allows for easy rinsing.
  • Dilutable for various cleaning tasks.
  • Comes in a large 55-gallon drum.