Learn More
Med Vet International Cleaner Disinfectant Cavicide Btl 24oz
Catalog No. 502091727
Encompass_Preferred
CaviCide is a convenient, ready-to-use, intermediate-level surface disinfectant which is effective against TB, HBV, HCV, viruses (hydrophilic and lipophilic), bacteria (including MRSA and VRE) and fungi. It is safe for all areas of the facility including NICU, operating rooms, isolation rooms, patient care areas and laboratories. When used as directed, it will also effectively clean and decontaminate critical and semi-critical instrumentation. CaviCide is safe for use on non-porous surfaces. It is perfect for cleaning and decontamination of environmental and medical device surfaces.
Catalog No. 50-209-1727 Supplier Med Vet International Supplier No. MAP131024HH
CaviCide is a convenient, ready-to-use, intermediate-level surface disinfectant which is effective against TB, HBV, HCV, viruses (hydrophilic and lipophilic), bacteria (including MRSA and VRE) and fungi. It is safe for all areas of the facility including NICU, operating rooms, isolation rooms, patient care areas and laboratories. When used as directed, it will also effectively clean and decontaminate critical and semi-critical instrumentation. CaviCide is safe for use on non-porous surfaces. It is perfect for cleaning and decontamination of environmental and medical device surfaces.