Learn More
Essendant Dishwashing Liquid, Lemon Scent, 38 oz Bottle, 8/Carton
Catalog No. 2680073
Encompass_Preferred
Joy Professional Manual Pot & Pan Dish Detergent cleans a wide variety of soils and leave your dishes with a fresh lemon scent. Forms surfactant micelles that stays suspended in wash water to provide micro-reservoirs of reserve cleaning power. A rich layer of suds provides high mileage performance for your dishes. Commercial grade detergent. Concentrated: Uses one to three fluid ounces of detergent per ten-gallon sink.
Catalog No. 26-800-73 Supplier Essendant Supplier No. JOY43606CT
Joy Professional Manual Pot & Pan Dish Detergent cleans a wide variety of soils and leave your dishes with a fresh lemon scent. Forms surfactant micelles that stays suspended in wash water to provide micro-reservoirs of reserve cleaning power. A rich layer of suds provides high mileage performance for your dishes. Commercial grade detergent. Concentrated: Uses one to three fluid ounces of detergent per ten-gallon sink.