Learn More
MSC Rochester Midland Corporation Hand Sanitizer, Foam, Dispenser Refill, 1 L, Fragrance Free
Catalog No. 502838307
Encompass
This foaming hand sanitizer cleans and sanitizes simultaneously, containing 0.13% Benzalkonium Chloride to kill 99.99% of common germs within 15 seconds. It is a 1 L dispenser refill, formulated without added dye or fragrance, making it suitable for various facilities, including those in food-related industries.
  • Fast-acting formula cleans and sanitizes at the same time
  • Kills 99.99% of common germs within 15 seconds
  • No added dye or fragrance
  • Ideal for use in food related facilities
  • Must be rinsed off after use
  • Contains a strong antibacterial agent with no iodine
  • Complies with E2 chlorine equivalency testing
  • Accepted for use in federally inspected meat and poultry plants
  • Biodegradable formulation
Catalog No. 50-283-8307 Supplier MSC Supplier No. 54472238
This foaming hand sanitizer cleans and sanitizes simultaneously, containing 0.13% Benzalkonium Chloride to kill 99.99% of common germs within 15 seconds. It is a 1 L dispenser refill, formulated without added dye or fragrance, making it suitable for various facilities, including those in food-related industries.
  • Fast-acting formula cleans and sanitizes at the same time
  • Kills 99.99% of common germs within 15 seconds
  • No added dye or fragrance
  • Ideal for use in food related facilities
  • Must be rinsed off after use
  • Contains a strong antibacterial agent with no iodine
  • Complies with E2 chlorine equivalency testing
  • Accepted for use in federally inspected meat and poultry plants
  • Biodegradable formulation