Learn More
Essendant No Rinse Sanitizer Concentrate, 1 gal Bottle, 4/Carton
Catalog No. 2678978
Encompass_Preferred
Meets food code requirements as a no rinse sanitizer and is perfect for a 3-bin sink and clean-in-place applications. Economical, concentrated formula that makes up to 512 gallons (1:512) and can be used as a disinfectant at 1:128. Use on floors, walls, furniture, bathtubs, shower stalls, toilet seats, food service and processing areas.
Catalog No. 26-789-78 Supplier Essendant Supplier No. RAC74389
Meets food code requirements as a no rinse sanitizer and is perfect for a 3-bin sink and clean-in-place applications. Economical, concentrated formula that makes up to 512 gallons (1:512) and can be used as a disinfectant at 1:128. Use on floors, walls, furniture, bathtubs, shower stalls, toilet seats, food service and processing areas.