Learn More
Electron Microscopy Sciences Cleaning Basket, Stainless Steel
SDP
Catalog No. 50996740
Encompass_Preferred
Made from stainless steel. Keeps small parts together when cleaning ultrasonically. A spring handle applies firm pressure to the basket to keep it closed when in use. Its overall length is 6". Spherical basket has a 1 1/2" (38 mm) diameter.
Catalog No. 50-996-740 Supplier Electron Microscopy Sciences Supplier No. 62110
Made from stainless steel. Keeps small parts together when cleaning ultrasonically. A spring handle applies firm pressure to the basket to keep it closed when in use. Its overall length is 6". Spherical basket has a 1 1/2" (38 mm) diameter.