Learn More
Cole-Parmer Round Sieve Brush for 100 Mesh Sieves and Coarser, Brass Bristles
Catalog No. NC0274914 Shop All Cole-Parmer Products
Encompass
These round sieve brushes are ideal for cleaning fine and coarse mesh sieves. The nylon brush is to clean 120 mesh and finer while brass bristle brushes are more suited for 100 mesh and coarser. Proper maintenance and cleaning of sieves prevents damage to sieve openings, keeping them clean and free of particles and ensures longest possible in-spec service life and accurate results.Each brush measures 6” long and the diameter of the bristle head is 2”.
This item is not returnable. View return policy
Catalog No. NC0274914 Supplier Cole-Parmer Supplier No. 5998697
These round sieve brushes are ideal for cleaning fine and coarse mesh sieves. The nylon brush is to clean 120 mesh and finer while brass bristle brushes are more suited for 100 mesh and coarser. Proper maintenance and cleaning of sieves prevents damage to sieve openings, keeping them clean and free of particles and ensures longest possible in-spec service life and accurate results.Each brush measures 6” long and the diameter of the bristle head is 2”.