Learn More
STERIS CORPORATION CLEANING BRUSH 12IN/2MM 3PK
Supplier:  STERIS CORPORATION PK45542
Encompass
Brush; Cleaning; Nylon Twisted Wire; 2 dia. x 12 in. L; Color bead: Red; Use On: Laparoscopic instruments, suction devices; White medical grade nylon bristles with surgical stainless steel twisted and spun wire; Reusable;
Catalog No. NC9387806