Learn More
BURKLE INC Cleaning brush, length 39.37 in. (100 cm), Ø 0.98 in. (25 mm), PVC/V2A
Catalog No. 5000025964
Encompass_Preferred
Cleaning brushes provide effective and thorough cleaning in various areas. Made of high-quality stainless steel and equipped with hard-wearing bristles, they effortlessly remove dirt, deposits and impurities from surfaces. They are ideal for cleaning samplers and narrow containers.
Catalog No. 50-000-25964 Supplier BURKLE INC Supplier No. 53040100
Cleaning brushes provide effective and thorough cleaning in various areas. Made of high-quality stainless steel and equipped with hard-wearing bristles, they effortlessly remove dirt, deposits and impurities from surfaces. They are ideal for cleaning samplers and narrow containers.