Learn More
Chemglass Life Sciences Cleaning Brush, 7/8" Dia x 16" Long
SDP
Supplier:  Chemglass Life Sciences CG114501
Encompass_Preferred
Cleaning Brush, 7/8" Dia x 16" Long. Soft sponge foam brush cleans glassware without absorbing water. Foam bristles never become soggy or limp. Will also not scratch or chip fragile glass items. Also will last up to 10 times longer than normal nylon brushes and up to 20 times longer than sponges. Sold as a set or individually.
Catalog No. 50-871-989