Learn More
Chemglass Life Sciences Cleaning Brush 9" long
SDP
Catalog No. 50871993
Encompass_Preferred
Cleaning Brush 9" long. Brush has a 3" wide x 4-1/2" rectangular non-abrasive scrubbing pad at the end of either a 9" or 33" handle for cleaning larger cylindrical reactors. Chemically resistant brush head will not scratch or mar delicate or coated surfaces.
Catalog No. 50-871-993 Supplier Chemglass Life Sciences Supplier No. CG114601
Cleaning Brush 9" long. Brush has a 3" wide x 4-1/2" rectangular non-abrasive scrubbing pad at the end of either a 9" or 33" handle for cleaning larger cylindrical reactors. Chemically resistant brush head will not scratch or mar delicate or coated surfaces.