Learn More
Chemglass Life Sciences Cleaning Brush Set; Contains 1 Each of the Above
SDP
Catalog No. 50871992
Encompass_Preferred
Cleaning Brush Set; Contains 1 Each of the Above. Soft sponge foam brush cleans glassware without absorbing water. Foam bristles never become soggy or limp. Will also not scratch or chip fragile glass items. Also will last up to 10 times longer than normal nylon brushes and up to 20 times longer than sponges. Sold as a set or individually.
Catalog No. 50-871-992 Supplier Chemglass Life Sciences Supplier No. CG114504
Cleaning Brush Set; Contains 1 Each of the Above. Soft sponge foam brush cleans glassware without absorbing water. Foam bristles never become soggy or limp. Will also not scratch or chip fragile glass items. Also will last up to 10 times longer than normal nylon brushes and up to 20 times longer than sponges. Sold as a set or individually.