Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Cleaning and Disinfecting Kit
SDP

Catalog No. 2573648 Shop All Mopec Products
Change view
Click to view available options
pH:
Neutral
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
25-736-48 Neutral
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25-736-48 Supplier Mopec™ Supplier No. BE125
  • $465.00
    $424.65 / Each of 1

    Save $40.35

    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Cleaning and Disinfecting Kit prioritizes the health and safety of those in the pathology, mortuary and animal science industries.

Kit contains:

  • 32 oz. bottle of SaniPath Foaming Spray (Path Cloud)
  • 32 oz. bottle of EazySoak
  • 32 oz. bottle of SaniPath Liquid Spray (Bench Spray & Wipe)
  • 32 oz. bottle of Stainless Steel Cleaner
  • 3 pack of nylon brushes
  • 3 pack of stainless steel brushes
  • Heavy-duty caddy

Specifications

Product Type Cleaner
Color-coded No
Form Liquid
pH Neutral
Scent Unscented
Sterility Non-sterile
Includes 32 oz. Bottle of SaniPath Foaming Spray (Path Cloud), 32 oz. Bottle of EazySoak, 32 oz. Bottle of SaniPath Liquid Spray (Bench Spray & Wipe), 32 oz. Bottle of Stainless Steel Cleaner, 3 Nylon Brushes, 3 Stainless Steel Brushes, Heavy Duty Caddy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.