Learn More
MSC Schaefer Brush Cleaning and Finishing Brush, Synthetic Bristles, 1 ea
Catalog No. 502867510
Encompass
This cleaning and finishing brush is equipped with synthetic bristles and a durable wood handle, making it suitable for a variety of surface preparation tasks. It is specifically designed for applications on materials harder than brass.
  • Brush type: Cleaning and finishing
  • Bristle material: Synthetic
  • Block material: Wood
  • Application: Surfaces harder than brass
  • Brush length: 4-1/4 in
  • Bristle length: 0.7500 in
  • Brush width: 2-1/2 in
  • Overall length: 4-1/4 in
Catalog No. 50-286-7510 Supplier MSC Supplier No. 09305723
This cleaning and finishing brush is equipped with synthetic bristles and a durable wood handle, making it suitable for a variety of surface preparation tasks. It is specifically designed for applications on materials harder than brass.
  • Brush type: Cleaning and finishing
  • Bristle material: Synthetic
  • Block material: Wood
  • Application: Surfaces harder than brass
  • Brush length: 4-1/4 in
  • Bristle length: 0.7500 in
  • Brush width: 2-1/2 in
  • Overall length: 4-1/4 in